• USA Home
  • 8.07010 - 2-Phenylpropionaldehyde

EMAIL THIS PAGE TO A FRIEND
8.07010 EMD Millipore

2-Phenylpropionaldehyde

2-Phenylpropionaldehyde for synthesis. CAS No. 93-53-8, EC Number 202-255-5., for synthesis

Synonym: 2-Phenylpropanal, Hydratropaldehyde

  • CAS Number 93-53-8

  • Linear Formula C₉H₁₀O

  • Molecular Weight 134.17 g/mol

  •  EC Number 202-255-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8070100005)
bp   205 °C (1013 hPa)
vapor pressure   0.4 hPa (20 °C)
solubility   3.4 g/l
density   1.01 g/cm3 (20 °C)
flash point   79 °C
autoignition temp.   405 °C
mp   <-60 °C
LD 50 oral   LD50 Rat 2800 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.002 - 1.006
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?