• USA Home
  • 8.07012 - 1-Phenyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
8.07012 EMD Millipore

1-Phenyl-1-propanol

1-Phenyl-1-propanol for synthesis. CAS 93-54-9, chemical formula C₆H₅CH(OH)CH₂CH₃., for synthesis

Synonym: α-Phenylpropyl alcohol, Phenyl ethyl carbinol, Ethyl phenyl carbinol, α-Ethylbenzyl alcohol

  • CAS Number 93-54-9

  • Linear Formula C₉H₁₂O

  • Molecular Weight 136.19 g/mol

  •  EC Number 202-256-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Plastic bottle (8070120100)
bp   217 - 219 °C (1013 hPa)
vapor pressure   0.2 hPa (20 °C)
solubility   0.62 g/l
density   0.99 g/cm3 (20 °C)
flash point   95 °C
LD 50 oral   LD50 Rat 1500 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.992 - 0.994
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
RTECS 
DO5470000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?