• USA Home
  • 8.07014 - 3-Phenyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
8.07014 EMD Millipore

3-Phenyl-1-propanol

3-Phenyl-1-propanol for synthesis. CAS No. 122-97-4, EC Number 204-587-6., for synthesis

Synonym: γ-Phenylpropyl alcohol, Hydrocinnamyl alcohol

  • CAS Number 122-97-4

  • Linear Formula C₉H₁₂O

  • Molecular Weight 136.19 g/mol

  •  EC Number 204-587-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8070140005)
vapor pressure   0.5 hPa (20 °C)
solubility   0.1 g/l insoluble
density   1.00 g/cm3 (20 °C)
flash point   120 °C
autoignition temp.   430 °C
mp   -18 °C
LD 50 oral   LD50 Rat 2300 mg/kg
LD 50 dermal   LD50 Rabbit 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.000 - 1.002
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?