• USA Home
  • 8.07025 - 4-Isopropylbenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.07025 EMD Millipore

4-Isopropylbenzaldehyde

4-Isopropylbenzaldehyde for synthesis. CAS 122-03-2, chemical formula 4-(CH₃)₂CHC₆H₄CHO., for synthesis

Synonym: Cuminaldehyde

  • CAS Number 122-03-2

  • Linear Formula C₁₀H₁₂O

  • Molecular Weight 148.21 g/mol

  •  EC Number 204-516-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8070250100)
bp   232 - 235 °C (1013 hPa)
density   0.98 g/cm3 (20 °C)
flash point   93 °C
LD 50 oral   LD50 Rat 1390 mg/kg
LD 50 dermal   LD50 Rabbit 2800 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 90.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.978 - 0.981
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 29 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?