EMAIL THIS PAGE TO A FRIEND
8.07029 EMD Millipore

2-Pyridylmethanol

2-Pyridylmethanol for synthesis. CAS No. 586-98-1, EC Number 209-592-7., for synthesis

Synonym: 2-(Hydroxymethyl)pyridine, 2-Pyridylcarbinol

  • CAS Number 586-98-1

  • Linear Formula C₆H₇NO

  • Molecular Weight 109.12 g/mol

  •  EC Number 209-592-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   220 °C (1013 hPa)
vapor pressure   0.04 hPa (20 °C)
density   1.13 g/cm3 (20 °C)
flash point   117 °C
mp   -6 °C
pH   6 - 8 (100 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.5 % (a/a)
  Density (d 20 °C/ 4 °C): 1.131 - 1.133
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?