EMAIL THIS PAGE TO A FRIEND
8.07048 EMD Millipore

3-Methylpyridine

3-Methylpyridine for synthesis. CAS No. 108-99-6, EC Number 203-636-9., for synthesis

Synonym: 3-Picoline

  • CAS Number 108-99-6

  • Linear Formula C₆H₇N

  • Molecular Weight 93.13 g/mol

  •  EC Number 203-636-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8070480250)
bp   144 °C (1013 hPa)
expl. lim.     1.3 - 8.7 %(V)
vapor pressure   6 hPa
density   0.96 g/cm3 (20 °C)
flash point   40 °C
mp   -18 °C
pH   10 (100 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 400 - 800 mg/kg
LD 50 dermal   LD50 Rabbit 200 - 1000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.955 - 0.957
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302 + H332: Harmful if swallowed or if inhaled
H311: Toxic in contact with skin.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
TJ5000000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2313 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2313 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2313 , 3, III
Categories of danger 
flammable, harmful, irritant
R Phrase 
R 10-20/21/22-36/37/38
Flammable.Harmful by inhalation, in contact with skin and if swallowed.Irritating to eyes, respiratory system and skin.
S Phrase 
S 26-36/37
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing and gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?