EMAIL THIS PAGE TO A FRIEND
8.07055 EMD Millipore

Oxindole

Oxindole for synthesis. CAS 59-48-3, molar mass 133.15 g/mol., for synthesis

Synonym: 2-Indolinone

  • CAS Number 59-48-3

  • Linear Formula C₈H₇NO

  • Molecular Weight 133.15 g/mol

  •  EC Number 200-429-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8070550010)
solubility   9.1 g/l
mp   122 - 125 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (HPLC): ≥ 98.0 % (a/a)
  Identity (IR-spectrum): passes test
  Melting range (lower value): 122 - 127
  Melting range (upper value): 122 - 127

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 79 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?