• USA Home
  • 8.07057 - Ethyl 2-pyridinecarboxylate

EMAIL THIS PAGE TO A FRIEND
8.07057 EMD Millipore

Ethyl 2-pyridinecarboxylate

Ethyl 2-pyridinecarboxylate for synthesis. CAS 2524-52-9, chemical formula 2-(C₂H₅OOC)C₅H₄N., for synthesis

Synonym: 2-Pyridinecarboxylic acid ethyl ester, Picolinic acid ethyl ester, Ethyl 2-picolinate

  • CAS Number 2524-52-9

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 219-758-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8070570025)
density   1.12 g/cm3 (25 °C)
flash point   107 °C
mp   1 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.118 - 1.122
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?