EMAIL THIS PAGE TO A FRIEND
8.07067 EMD Millipore

Phthalide

Phthalide for synthesis. CAS 87-41-2, molar mass 134.14 g/mol., for synthesis

Synonym: 1,3-Dihydrobenzo[c]furan-1-one, Oxophthalane, 1(3H)-Isobenzofuranone

  • CAS Number 87-41-2

  • Linear Formula C₈H₆O₂

  • Molecular Weight 134.14 g/mol

  •  EC Number 201-744-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8070670100)
vapor pressure   0.003 hPa (25 °C)
density   1.41 g/cm3 (18 °C)
autoignition temp.   455 °C
mp   70 - 75 °C
bulk density   600 kg/m3
LD 50 oral   LD50 Rat 5600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 70 °C
  Melting range (upper value): ≤ 73 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 20 90
RTECS 
TI3520000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?