• USA Home
  • 8.07070 - DL-α-Aminophenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.07070 EMD Millipore

DL-α-Aminophenylacetic acid

for synthesis

Synonym: DL-α-Phenylaminoacetic acid, DL-α-Phenylglycine

  • CAS Number 2835-06-5

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 220-608-1

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8070700100)
mp   290 °C (sublimed)
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?