• USA Home
  • 8.07073 - (R)-(-)-α-Aminophenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.07073 EMD Millipore

(R)-(-)-α-Aminophenylacetic acid

(R)-(-)-α-Aminophenylacetic acid for resolution of racemates for synthesis. CAS 875-74-1, chemical formula C₆H₅CH(NH₂)COOH., for resolution of racemates for synthesis

Synonym: (R)-(-)-α-Phenylaminoacetic acid, D(-)-α-Phenylglycine

  • CAS Number 875-74-1

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 212-876-3

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8070730025)
density   1.2 g/cm3 (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Spec. rotation [α²0/D (c=2 in HCl 1 mol/l): -158.0 - -156.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?