• USA Home
  • 8.07105 - Palladium/barium sulfate (5% Pd)

EMAIL THIS PAGE TO A FRIEND
8.07105 EMD Millipore

Palladium/barium sulfate (5% Pd)

hydrogenation catalyst (oxidic form) for synthesis

Purchase

Properties

packaging   10 g in Glass bottle (8071050010)
pH   2.5 - 5 (10 g/l, H₂O, 20 °C) (slurry)
bulk density   700 - 900 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ICP-OES): 4.0 - 6.0 %
  Suitable for hydrogenation catalyst: passes test
  Identity (ICP-OES): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
3815 12 00
Storage class 
10 - 13 Other liquids and solids
WGK 
NWG not water endangering
Disposal 
27
Residues containing valuable recoverable metals should be forwarded for recycling. Container H.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?