• USA Home
  • 8.07113 - (S)-(-)-N-(1-Phenylethyl)succinamic acid

EMAIL THIS PAGE TO A FRIEND
8.07113 EMD Millipore

(S)-(-)-N-(1-Phenylethyl)succinamic acid

(S)-(-)-N-(1-Phenylethyl)succinamic acid for resolution of racemates for synthesis. CAS 21752-34-1, chemical formula C₆H₅CH(CH₃)NHCOCH₂CH₂COOH., for resolution of racemates for synthesis

Synonym: (-)-N-(1-Phenylethyl)succinic acid monoamide

  • CAS Number 21752-34-1

  • Linear Formula C₁₂H₁₅NO₃

  • Molecular Weight 221.26 g/mol

  •  EC Number 244-569-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   99 - 102 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 99 °C
  Melting range (upper value): ≤ 102 °C
  Spec. rotation [α²0/D (c=2 in ethanol): -112.0 - -109.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?