• USA Home
  • 8.07114 - Pentadecafluorooctanoic acid

EMAIL THIS PAGE TO A FRIEND
8.07114 EMD Millipore

Pentadecafluorooctanoic acid

Pentadecafluorooctanoic acid for synthesis. CAS 335-67-1, EC Number 206-397-9, chemical formula CF₃(CF₂)₆COOH., for synthesis

Synonym: Perfluorooctanoic acid

  • CAS Number 335-67-1

  • Linear Formula C₈HF₁₅O₂

  • Molecular Weight 414.06 g/mol

  •  EC Number 206-397-9

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Synthetic Reagents
packaging   10 g in Plastic bottle (8071140010)
solubility   3.4 g/l
density   1.8 g/cm3 (20 °C)
mp   45 - 50 °C
pH   2.6 (1 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric, calc. on anhydrous substance): ≥ 97.0 %
  Water (K. F.): ≤ 5.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H360D: May damage the unborn child.
H302 + H332: Harmful if swallowed or if inhaled
H318: Causes serious eye damage.
H351: Suspected of causing cancer.
H362: May cause harm to breast-fed children.
H372: Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement(s) 
P201: Obtain special instructions before use.
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P314: Get medical advice/ attention if you feel unwell.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
toxic for reproduction, corrosive, toxic, carcinogenic, irritant
R Phrase 
R 61-20/22-48/23-48/21/22-40-41-64
May cause harm to the unborn child.Also harmful by inhalation and if swallowed.Also toxic: danger of serious damage to health by prolonged exposure through inhalation.Also harmful: danger of serious damage to health by prolonged exposure in contact with skin and if swallowed.Limited evidence of a carcinogenic effect.Risk of serious damage to eyes.May cause harm to breastfed babies.
S Phrase 
S 53-45
Avoid exposure - obtain special instructions before use.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?