• USA Home
  • 8.07286 - Ethyl phenylacetate

EMAIL THIS PAGE TO A FRIEND
8.07286 EMD Millipore

Ethyl phenylacetate

Ethyl phenylacetate for synthesis. CAS 101-97-3, EC Number 202-993-8, chemical formula C₆H₅CH₂COOC₂H₅., for synthesis

Synonym: Phenylacetic acid ethyl ester

  • CAS Number 101-97-3

  • Linear Formula C₁₀H₁₂O₂

  • Molecular Weight 164.2 g/mol

  •  EC Number 202-993-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   500 ml in Glass bottle (8072860500)
bp   226 - 229 °C (1013 hPa)
vapor pressure   31 hPa (120 °C)
density   1.03 g/cm3 (20 °C)
flash point   97 °C
mp   -29 °C
LD 50 oral   LD50 Rat 3300 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 10
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?