• USA Home
  • 8.07292 - 3,5-Dihydro-4-phenyl-4H-1,2,4-triazole-3,5-dione

EMAIL THIS PAGE TO A FRIEND
8.07292 EMD Millipore

3,5-Dihydro-4-phenyl-4H-1,2,4-triazole-3,5-dione

3,5-Dihydro-4-phenyl-4H-1,2,4-triazole-3,5-dione for synthesis. CAS 4233-33-4, molar mass 175.14 g/mol., for synthesis

Synonym: 4-Phenyl-3,5-dihydro-4H-1,2,4-triazole-3,5-dione, PTAD

  • CAS Number 4233-33-4

  • Linear Formula C₈H₅N₃O₂

  • Molecular Weight 175.14 g/mol

  •  EC Number 224-191-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8072920001)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay at N (Elemental analysis): ≥ 95.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?