• USA Home
  • 8.07302 - Phthalimide potassium salt

EMAIL THIS PAGE TO A FRIEND
8.07302 EMD Millipore

Phthalimide potassium salt

Phthalimide potassium salt for synthesis. CAS 1074-82-4, molar mass 185.22 g/mol., for synthesis

Synonym: Potassium phthalimide

  • CAS Number 1074-82-4

  • Linear Formula C₈H₄KNO₂

  • Molecular Weight 185.22 g/mol

  •  EC Number 214-046-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8073020100)
density   1.63 g/cm3
mp   300 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2925 19 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?