EMAIL THIS PAGE TO A FRIEND
8.07333 EMD Millipore

n-Pentylamine

n-Pentylamine for synthesis. CAS 110-58-7, EC Number 203-780-2, chemical formula CH₃(CH₂)₄NH₂., for synthesis

Synonym: 1-Aminopentane, n-Amylamine

  • CAS Number 110-58-7

  • Linear Formula C₅H₁₃N

  • Molecular Weight 87.16 g/mol

  •  EC Number 203-780-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8073330025)
bp   103 °C (1013 hPa)
expl. lim.     2.2 - 22 %(V)
vapor pressure   120.9 hPa (47 °C)
density   0.75 g/cm3 (20 °C)
flash point   7 °C
autoignition temp.   305 °C
mp   -50 °C
LD 50 oral   LD50 Rat 470 mg/kg
LD 50 dermal   LD50 Rabbit 1120 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.753 - 0.757
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 19 99
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H302 + H312: Harmful if swallowed or in contact with skin
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P233: Keep container tightly closed.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
P403 + P235: Store in a well-ventilated place. Keep cool.
RTECS 
SC0300000
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1106 , 3 (8), II
Declaration (transport by air) IATA-DGR 
UN 1106 , 3 (8), II
Declaration (transport by sea) IMDG-Code 
UN 1106 , 3 (8), II
Categories of danger 
highly flammable, harmful, corrosive
R Phrase 
R 11-21/22-34
Highly flammable.Harmful in contact with skin and if swallowed.Causes burns.
S Phrase 
S 16-26-36/37/39-45
Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?