• USA Home
  • 8.07335 - 2-Phenylethyl bromide

EMAIL THIS PAGE TO A FRIEND
8.07335 EMD Millipore

2-Phenylethyl bromide

2-Phenylethyl bromide for synthesis. CAS No. 103-63-9, EC Number 203-130-8., for synthesis

Synonym: Phenethyl bromide, (2-Bromoethyl)benzene, 1-Bromo-2-phenylethane

  • CAS Number 103-63-9

  • Linear Formula C₈H₉Br

  • Molecular Weight 185.06 g/mol

  •  EC Number 203-130-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8073350250)
bp   220 °C (1013 hPa)
vapor pressure   0.3 hPa (25 °C)
solubility   0.039 g/l
density   1.37 g/cm3 (20 °C)
flash point   89 °C
mp   -56 °C
refractive index   1.5563 (20 °C, 589 nm)
LD 50 oral   LD50 Rat 811 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.5 % (a/a)
  Density (d 20 °C/ 4 °C): 1.366 - 1.369
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H302: Harmful if swallowed.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-36
Harmful if swallowed.Irritating to eyes.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?