• USA Home
  • 8.07470 - 2-Pyridinecarbaldehyde

EMAIL THIS PAGE TO A FRIEND
8.07470 EMD Millipore

2-Pyridinecarbaldehyde

2-Pyridinecarbaldehyde for synthesis. CAS No. 1121-60-4, EC Number 214-333-6., for synthesis

Synonym: Pyridine-2-aldehyde

  • CAS Number 1121-60-4

  • Linear Formula C₆H₅NO

  • Molecular Weight 107.11 g/mol

  •  EC Number 214-333-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8074700250)
bp   181 °C (1013 hPa)
vapor pressure   1.2 hPa (20 °C)
density   1.12 g/cm3 (20 °C)
flash point   73 °C
autoignition temp.   235 °C
mp   -21 °C
pH   6.5 (111 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 585 mg/kg
LD 50 dermal   LD50 Rat > 2000 mg/kg
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.120 - 1.122
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
H331: Toxic if inhaled.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
Categories of danger 
toxic, irritant, sensitizing, dangerous for the environment
R Phrase 
R 22-23-36/37/38-43-51/53
Harmful if swallowed.Toxic by inhalation.Irritating to eyes, respiratory system and skin.May cause sensitisation by skin contact.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24-37-45-61
Avoid contact with skin.Wear suitable gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?