EMAIL THIS PAGE TO A FRIEND
8.07504 EMD Millipore

2-Phenylimidazole

2-Phenylimidazole for synthesis. CAS 670-96-2, molar mass 144.17 g/mol., for synthesis

  • CAS Number 670-96-2

  • Linear Formula C₉H₈N₂

  • Molecular Weight 144.17 g/mol

  •  EC Number 211-581-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8075040100)
bp   340 °C (1013 hPa)
vapor pressure   26 hPa (216 °C)
density   0.6 g/cm3
flash point   200 °C
mp   146 - 150 °C
bulk density   500 kg/m3
LD 50 oral   LD50 Rat 864 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 146 °C
  Melting range (upper value): ≤ 150 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Hazard Statement(s) 
H302: Harmful if swallowed.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?