• USA Home
  • 8.07508 - N-(2-Chloroethyl)-piperidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.07508 EMD Millipore

N-(2-Chloroethyl)-piperidine hydrochloride

N-(2-Chloroethyl)-piperidine hydrochloride for synthesis. CAS 2008-75-5, molar mass 184.1 g/mol., for synthesis

Synonym: 2-Piperidinoethyl chloride hydrochloride, N-(2-Chloroethyl)-piperidinium chloride

  • CAS Number 2008-75-5

  • Linear Formula C₇H₁₅Cl₂N

  • Molecular Weight 184.1 g/mol

  •  EC Number 217-920-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8075080100)
autoignition temp.   >350 °C
mp   234 - 237 °C
LD 50 oral   LD50 Rat 17 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 228 °C
  Melting range (upper value): ≤ 234 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Hazard Statement(s) 
H300: Fatal if swallowed.
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Very toxic  Very toxic
Declaration (railroad and road) ADR, RID 
UN 2928 , 6.1 (8), II
Declaration (transport by air) IATA-DGR 
UN 2928 , 6.1 (8), II
Declaration (transport by sea) IMDG-Code 
UN 2928 , 6.1 (8), II
Categories of danger 
very toxic, corrosive
R Phrase 
R 28-34
Very toxic if swallowed.Causes burns.
S Phrase 
S 26-28-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.After contact with skin, wash immediately with plenty of soap and water.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?