• USA Home
  • 8.07511 - 2-Phenylethyl chloride

EMAIL THIS PAGE TO A FRIEND
8.07511 EMD Millipore

2-Phenylethyl chloride

2-Phenylethyl chloride for synthesis. CAS No. 622-24-2, EC Number 210-725-6., for synthesis

Synonym: Phenethyl chloride, (2-Chloroethyl)benzene

  • CAS Number 622-24-2

  • Linear Formula C₈H₉Cl

  • Molecular Weight 140.61 g/mol

  •  EC Number 210-725-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8075110100)
bp   82 - 84 °C (21 hPa)
density   1.07 g/cm3 (20 °C)
flash point   64 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.071 - 1.072
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?