EMAIL THIS PAGE TO A FRIEND
8.07567 EMD Millipore

N-Phenylurea

N-Phenylurea for synthesis. CAS 64-10-8, EC Number 200-576-5, chemical formula C₆H₅NHCONH₂., for synthesis

Synonym: Phenylcarbamide

  • CAS Number 64-10-8

  • Linear Formula C₇H₈N₂O

  • Molecular Weight 136.15 g/mol

  •  EC Number 200-576-5

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks,
packaging   5 g in Glass bottle (8075670005)
solubility   10 g/l
mp   144 - 147 °C
LD 50 oral   LD50 Rat 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 144 °C
  Melting range (upper value): ≤ 147 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 21 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?