• USA Home
  • 8.07595 - Resorcinol dimethyl ether

EMAIL THIS PAGE TO A FRIEND
8.07595 EMD Millipore

Resorcinol dimethyl ether

Resorcinol dimethyl ether for synthesis. CAS 151-10-0, EC Number 205-783-4, chemical formula C₆H₄-1,3-(OCH₃)₂., for synthesis

Synonym: 1,3-Dimethoxybenzene

  • CAS Number 151-10-0

  • Linear Formula C₈H₁₀O₂

  • Molecular Weight 138.16 g/mol

  •  EC Number 205-783-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8075950050)
bp   217 - 218 °C
density   1.07 g/cm3 (20 °C)
flash point   88 °C
mp   -52 °C
refractive index   1.524 (20 °C, 589 nm)
LD 50 oral   LD50 Rat 2829 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.067 - 1.069
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?