EMAIL THIS PAGE TO A FRIEND
8.08385 EMD Millipore

Tropine

Tropine for synthesis. CAS 120-29-6, pH 12 (100 g/l, H₂O, 20 °C)., for synthesis

Synonym: Tropan-3α-ol, 8-Aza-N-methylbicyclo[3.2.1.]octan-3-ol, 3-Tropanol

  • CAS Number 120-29-6

  • Linear Formula C₈H₁₅NO

  • Molecular Weight 141.21 g/mol

  •  EC Number 204-384-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8083850025)
bp   232 - 234 °C (1013 hPa)
solubility   100 g/l
density   1.04 g/cm3 (20 °C)
mp   63 °C
pH   12 (100 g/l, H₂O, 20 °C)
bulk density   350 kg/m3
LD 50 oral   LD50 Rat 1050 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 62 °C
  Melting range (upper value): ≤ 65 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2939 80 00
Hazard Statement(s) 
H302 + H332: Harmful if swallowed or if inhaled
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 20/22
Harmful by inhalation and if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?