EMAIL THIS PAGE TO A FRIEND
8.08465 EMD Millipore

Ethyl undecanoate

Ethyl undecanoate for synthesis. CAS 627-90-7, EC Number 211-018-5, chemical formula CH₃(CH₂)₉COOC₂H₅., for synthesis

Synonym: Undecanoic acid ethyl ester

  • CAS Number 627-90-7

  • Linear Formula C₁₃H₂₆O₂

  • Molecular Weight 214.34 g/mol

  •  EC Number 211-018-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8084650100)
bp   104 - 106 °C (5 hPa)
density   0.86 g/cm3 (20 °C)
flash point   >100 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.862 - 0.864
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?