EMAIL THIS PAGE TO A FRIEND
8.08516 EMD Millipore

γ-Pentanolactone

γ-Pentanolactone for synthesis. CAS 108-29-2, pH 7 (H₂O, 20 °C)., for synthesis

Synonym: 4-Pentanolide, γ-Valerolactone

  • CAS Number 108-29-2

  • Linear Formula C₅H₈O₂

  • Molecular Weight 100.12 g/mol

  •  EC Number 203-569-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8085160100)
bp   205 - 206 °C (1013 hPa)
vapor pressure   1 hPa (20 °C)
density   1.05 g/cm3 (20 °C)
flash point   81 °C
mp   -31 °C
pH   7 (H₂O, 20 °C)
LD 50 oral   LD50 Rat 8800 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.053 - 1.055
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 20 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?