• USA Home
  • 8.08526 - 3,4-Dimethoxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.08526 EMD Millipore

3,4-Dimethoxybenzoic acid

3,4-Dimethoxybenzoic acid for synthesis. CAS No. 93-07-2, EC Number 202-215-7., for synthesis

Synonym: Veratric acid

  • CAS Number 93-07-2

  • Linear Formula C₉H₁₀O₄

  • Molecular Weight 182.17 g/mol

  •  EC Number 202-215-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8085260100)
solubility   0.5 g/l
mp   179 - 182 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
RTECS 
DG8598750
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?