• USA Home
  • 8.08541 - Ethyl isopentanoate

EMAIL THIS PAGE TO A FRIEND
8.08541 EMD Millipore

Ethyl isopentanoate

Ethyl isopentanoate for synthesis. CAS 108-64-5, EC Number 203-602-3, chemical formula (CH₃)₂CHCH₂COOC₂H₅., for synthesis

Synonym: Ethyl iso-pentanoate, Ethyl 3-methylbutanoate, 3-Methyl butyric ethyl ester

  • CAS Number 108-64-5

  • Linear Formula C₇H₁₄O₂

  • Molecular Weight 130.18 g/mol

  •  EC Number 203-602-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8085410250)
bp   133 - 135 °C (1013 hPa)
vapor pressure   11 hPa (25 °C)
solubility   2.00 g/l
density   0.86 g/cm3 (20 °C)
flash point   22 °C
mp   -99 °C
LD 50 oral   LD50 Rat > 5000 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.864 - 0.866
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 60 90
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P233: Keep container tightly closed.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 3272 , 3, III
Declaration (transport by air) IATA-DGR 
UN 3272 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 3272 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?