EMAIL THIS PAGE TO A FRIEND
8.08601 EMD Millipore

1-Tetradecene

1-Tetradecene for synthesis. CAS 1120-36-1, molar mass 196.37 g/mol., for synthesis

  • CAS Number 1120-36-1

  • Linear Formula C₁₄H₂₈

  • Molecular Weight 196.37 g/mol

  •  EC Number 214-306-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8086010100)
bp   251 °C (1013 hPa)
expl. lim.     0.5 - 5.4 %(V)
vapor pressure   0.013 hPa (25 °C)
density   0.77 g/cm3 (20 °C)
flash point   107 °C
autoignition temp.   235 °C
mp   -13 °C
viscosity kinematic   2.61 mm2/s (20 °C)
LD 50 oral   LD50 Rat > 10000 mg/kg
LD 50 dermal   LD50 Rabbit > 10000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 93.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.771 - 0.773
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2901 29 00
Hazard Statement(s) 
H304: May be fatal if swallowed and enters airways.
Precautionary Statement(s) 
P301 + P310: IF SWALLOWED: Immediately call a POISON CENTER/doctor.
P331: Do NOT induce vomiting.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 65
Harmful: may cause lung damage if swallowed.
S Phrase 
S 62
If swallowed, do not induce vomiting: seek medical advice immediately and show this container or label.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?