• USA Home
  • 8.08606 - 2,4,6-Trimethylaniline

EMAIL THIS PAGE TO A FRIEND
8.08606 EMD Millipore

2,4,6-Trimethylaniline

2,4,6-Trimethylaniline for synthesis. CAS 88-05-1, chemical formula 2,4,6-(CH₃)₃C₆H₂NH₂., for synthesis

Synonym: 2-Aminomesitylene, Mesidine

  • CAS Number 88-05-1

  • Linear Formula C₉H₁₃N

  • Molecular Weight 135.21 g/mol

  •  EC Number 201-794-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8086060100)
bp   232 - 234 °C (1013 hPa)
density   0.96 g/cm3 (20 °C)
flash point   96 °C
mp   -5 °C
LD 50 oral   LD50 Rat 743 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.962 - 0.964
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
BZ0700000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, irritant
R Phrase 
R 22-36
Harmful if swallowed.Irritating to eyes.
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?