• USA Home
  • 8.08613 - Trichloroacetyl isocyanate

EMAIL THIS PAGE TO A FRIEND
8.08613 EMD Millipore

Trichloroacetyl isocyanate

Trichloroacetyl isocyanate for synthesis. CAS 3019-71-4, EC Number 221-165-7, chemical formula CCl₃CONCO., for synthesis

  • CAS Number 3019-71-4

  • Linear Formula C₃Cl₃NO₂

  • Molecular Weight 188.39 g/mol

  •  EC Number 221-165-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8086130005)
bp   40 - 42 °C (20 hPa)
density   1.59 g/cm3 (20 °C)
flash point   65 °C
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.585 - 1.595
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2929 10 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P260: Do not breathe spray.
P262: Do not get in eyes, on skin, or on clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P342 + P311: If experiencing respiratory symptoms: Call a POISON CENTER/doctor.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
11
Organic acid halides, anhydrides and isocyanates can be added dropwise to an excess of methanol (Cat. No. 822283) to convert them into the corresponding methyl esters or methyl carbamates. If necessary, neutralize with sodium hydroxide solution (Cat. No. 105587). Fill into container A.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2206 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2206 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2206 , 6.1, II
Categories of danger 
irritant, sensitizing
R Phrase 
R 36/37/38-42
Irritating to eyes, respiratory system and skin.May cause sensitisation by inhalation.
S Phrase 
S 23-24-26-45
Do not breathe spray.Avoid contact with skin.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?