• USA Home
  • 8.08624 - 1,2,4,5-Tetramethylbenzene

EMAIL THIS PAGE TO A FRIEND
8.08624 EMD Millipore

1,2,4,5-Tetramethylbenzene

1,2,4,5-Tetramethylbenzene for synthesis. CAS 95-93-2, chemical formula C₆H₂-1,2,4,5-(CH₃)₄., for synthesis

Synonym: Durol, Durene

  • CAS Number 95-93-2

  • Linear Formula C₁₀H₁₄

  • Molecular Weight 134.22 g/mol

  •  EC Number 202-465-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8086240100)
bp   191 - 193 °C
vapor pressure   26 hPa (90 °C)
solubility   0.00348 g/l
flash point   74 °C
mp   78 - 81 °C
LD 50 oral   LD50 Rat 6700 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 76 °C
  Melting range (upper value): ≤ 80 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 90 00
Hazard Statement(s) 
H228: Flammable solid.
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P273: Avoid release to the environment.
Storage class 
4.1B Flammable solid hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 1325 , 4.1, II
Declaration (transport by air) IATA-DGR 
UN 1325 , 4.1, II
Declaration (transport by sea) IMDG-Code 
UN 1325 , 4.1, II
Categories of danger 
highly flammable, dangerous for the environment
R Phrase 
R 11-50/53
Highly flammable.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?