EMAIL THIS PAGE TO A FRIEND
8.08633 EMD Millipore

2-Tridecanone

2-Tridecanone for synthesis. CAS No. 593-08-8, EC Number 209-784-0., for synthesis

  • CAS Number 593-08-8

  • Linear Formula C₁₃H₂₆O

  • Molecular Weight 198.35 g/mol

  •  EC Number 209-784-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Plastic bottle (8086330250)
density   0.82 g/cm3
flash point   122 °C
mp   29 - 31 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 19 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?