• USA Home
  • 8.08638 - 3,4,5-Trimethoxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.08638 EMD Millipore

3,4,5-Trimethoxybenzoic acid

3,4,5-Trimethoxybenzoic acid for synthesis. CAS No. 118-41-2, EC Number 204-248-2., for synthesis

Synonym: Gallic acid trimethyl ether

  • CAS Number 118-41-2

  • Linear Formula C₁₀H₁₂O₅

  • Molecular Weight 212.20 g/mol

  •  EC Number 204-248-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Plastic bottle (8086380250)
bp   225 - 227 °C (13 hPa)
mp   169 - 171 °C
bulk density   270 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 167 °C
  Melting range (upper value): ≤ 170 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?