EMAIL THIS PAGE TO A FRIEND
8.08669 EMD Millipore

Xanthone

Xanthone for synthesis. CAS 90-47-1, molar mass 196.21 g/mol., for synthesis

Synonym: 9-Xanthenone

  • CAS Number 90-47-1

  • Linear Formula C₁₃H₈O₂

  • Molecular Weight 196.21 g/mol

  •  EC Number 201-997-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8086690025)
mp   172 - 174 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 173 °C
  Melting range (upper value): ≤ 176 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
RTECS 
ZD5711000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?