• USA Home
  • 8.08729 - Trimethyltin chloride

EMAIL THIS PAGE TO A FRIEND
8.08729 EMD Millipore

Trimethyltin chloride

Trimethyltin chloride for synthesis. CAS 1066-45-1, EC Number 213-917-8, chemical formula (CH₃)₃SnCl., for synthesis

Synonym: Trimethylchlorotin, Chlorotrimethylstannane, Chlorotrimethyltin

  • CAS Number 1066-45-1

  • Linear Formula C₃H₉ClSn

  • Molecular Weight 199.24 g/mol

  •  EC Number 213-917-8

Purchase

Properties

flash point   97 °C
mp   36 - 39 °C
LD 50 oral   LD50 Rat 12.6 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 36 °C
  Melting range (upper value): ≤ 39 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H300 + H310 + H330: Fatal if swallowed, in contact with skin or if inhaled
H410: Very toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
15
Heavy metal-containing solutions and solids: container E. Stir Raney nickel (also: Urushibara nickel) in the form of an aqueous suspension into hydrochloric acid (Cat. No. 100312) until dissolved (container E). Neither Raney nickel itself nor any filter residues should be allowed to dry out, otherwise they will spontaneously ignite in air. In this context, heavy metal means any compound of antimony, arsenic, cadmium, chromium(VI), copper, lead, nickel and tin, as well as these subtances in metallic form, if they are classified as hazardous (acc. to AbfallverzeichnisV - Waste Catalogue Ordinance, Appendix 3). Other heavy metals should be collected separately.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Very toxic  Very toxic
Declaration (railroad and road) ADR, RID 
UN 3146 , 6.1, I
Declaration (transport by air) IATA-DGR 
UN 3146 , 6.1, I
Declaration (transport by sea) IMDG-Code 
UN 3146 , 6.1, I, Marine Pollutant: P
Categories of danger 
very toxic, dangerous for the environment
R Phrase 
R 26/27/28-50/53
Very toxic by inhalation, in contact with skin and if swallowed.Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-27-28-45-60-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Take off immediately all contaminated clothing.After contact with skin, wash immediately with plenty of soap and water.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).This material and its container must be disposed of as hazardous waste.Avoid release to the environment. Refer to special instructions/ Safety data sheets.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?