• USA Home
  • 8.10165 - Trimethylsulfoxoniumiodide

EMAIL THIS PAGE TO A FRIEND
8.10165 EMD Millipore

Trimethylsulfoxoniumiodide

Trimethylsulfoxoniumiodide for synthesis. CAS 1774-47-6, EC Number 217-204-2, chemical formula (CH₃)₃SOI., for synthesis

  • CAS Number 1774-47-6

  • Linear Formula C₃H₉IOS

  • Molecular Weight 220.07 g/mol

  •  EC Number 217-204-2

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfoxides,
packaging   100 g in Glass bottle (8101650100)
solubility   15 g/l
autoignition temp.   200 °C
mp   165 - 175 °C decomposes
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?