• USA Home
  • 8.10489 - Palladium/calcium carbonate (5 % Pd)

EMAIL THIS PAGE TO A FRIEND
8.10489 EMD Millipore

Palladium/calcium carbonate (5 % Pd)

Lindlar hydrogenation catalyst (poisoned with lead) for synthesis

Synonym: Hydrogenation catalyst

Purchase

Properties

packaging   5 g in Glass bottle (8104890005)
bulk density   500 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (ICP-OES): 4.0 - 6.0 %
  Suitable for hydrogenation catalyst: passes test
  Identity (ICP-OES): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
3815 12 00
Hazard Statement(s) 
H360Df: May damage the unborn child. Suspected of damaging fertility.
H373: May cause damage to organs through prolonged or repeated exposure.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P201: Obtain special instructions before use.
P273: Avoid release to the environment.
P314: Get medical advice/ attention if you feel unwell.
Storage class 
6.1 D Non combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
WGK 
NWG not water endangering
Disposal 
27
Residues containing valuable recoverable metals should be forwarded for recycling. Container H.
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3077 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3077 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3077 , 9, III, Marine Pollutant: P, Segregation Group: 7 (Heavy metals and their salts (including their organometallic compounds))9 (Lead and its compounds)
Categories of danger 
toxic for reproduction, harmful, dangerous for the environment
R Phrase 
R 61-20/22-33-51/53-62
May cause harm to the unborn child.Also harmful by inhalation and if swallowed.Danger of cumulative effects.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.Possible risk of impaired fertility.
S Phrase 
S 53-36/37-45-61
Avoid exposure - obtain special instructions before use.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?