• USA Home
  • 8.10557 - Triethyl phosphonoacetate

EMAIL THIS PAGE TO A FRIEND
8.10557 EMD Millipore

Triethyl phosphonoacetate

Triethyl phosphonoacetate for synthesis. CAS 867-13-0, EC Number 212-757-6, chemical formula C₂H₅OCOCH₂PO(OC₂H₅)₂., for synthesis

Synonym: Triethyl phosphonoacetate, Ethoxycarbonylmethanephosphonic acid diethyl ester

  • CAS Number 867-13-0

  • Linear Formula C₈H₁₇O₅P

  • Molecular Weight 224.19 g/mol

  •  EC Number 212-757-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8105570100)
bp   260 - 262 °C (1013 hPa)
density   1.12 g/cm3 (20 °C)
flash point   105 °C
autoignition temp.   445 °C
mp   -24 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.121 - 1.123
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
AG9800000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?