• USA Home
  • 8.10569 - Nitrosyl tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
8.10569 EMD Millipore

Nitrosyl tetrafluoroborate

Nitrosyl tetrafluoroborate for synthesis. CAS 14635-75-7, EC Number 238-679-2, chemical formula NOBF₄., for synthesis

  • CAS Number 14635-75-7

  • Linear Formula BF₄NO

  • Molecular Weight 116.8 g/mol

  •  EC Number 238-679-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8105690005)
density   2.185 g/cm3
mp   157 - 160 °C (sublimed)
shipped in   ambient
storage temp.   Store at +2°C to +8°C.
specifications   Assay (acidimetric): ≥ 94.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2812 90 00
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
EUH031: Contact with acids liberates toxic gas.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
24
Liquid inorganic halides and hydrolysis-sensitive reagents can be carefully stirred dropwise into ice-cool 10% sodium hydroxide solution (Cat. No. 105587) (caution: caustic gases!); container D or E.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 3260 , 8, II
Declaration (transport by air) IATA-DGR 
UN 3260 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 3260 , 8, II, Segregation Group: 1 (Acids)
Categories of danger 
corrosive
R Phrase 
R 31-34
Contact with acids liberates toxic gas.Causes burns.
S Phrase 
S 7/8-26-36/37/39-45
Keep container tightly closed and dry.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?