• USA Home
  • 8.10591 - 2,4,6-Triisopropylbenzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
8.10591 EMD Millipore

2,4,6-Triisopropylbenzenesulfonyl chloride

2,4,6-Triisopropylbenzenesulfonyl chloride for synthesis. CAS 6553-96-4, chemical formula 2,4,6-(CH₃)₂CH₃C₆H₂SO₂Cl., for synthesis

Synonym: TPSCl

  • CAS Number 6553-96-4

  • Linear Formula C₁₅H₂₃ClO₂S

  • Molecular Weight 302.85 g/mol

  •  EC Number 229-479-6

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds More...
packaging   25 g in Glass bottle (8105910025)
solubility   <0.1 g/l (slow decomposition)
mp   93 - 96 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 93 °C
  Melting range (upper value): ≤ 96 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 99 00
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
11
Organic acid halides, anhydrides and isocyanates can be added dropwise to an excess of methanol (Cat. No. 822283) to convert them into the corresponding methyl esters or methyl carbamates. If necessary, neutralize with sodium hydroxide solution (Cat. No. 105587). Fill into container A.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 3261 , 8, III
Declaration (transport by air) IATA-DGR 
UN 3261 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 3261 , 8, III, Segregation Group: 1 (Acids)
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?