• USA Home
  • 8.13718 - Crown ether/Benzo-15-crown-5

EMAIL THIS PAGE TO A FRIEND
8.13718 EMD Millipore

Crown ether/Benzo-15-crown-5

Crown ether/Benzo-15-crown-5 for synthesis. CAS 14098-44-3, molar mass 268.3 g/mol., for synthesis

Synonym: Benzo-15-crown-5, 1,4,7,10,13-Pentaoxa[13]orthocyclophane

  • CAS Number 14098-44-3

  • Linear Formula C₁₄H₂₀O₅

  • Molecular Weight 268.3 g/mol

  •  EC Number 237-947-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8137180005)
mp   79 - 81 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 78 °C
  Melting range (upper value): ≤ 82 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?