• USA Home
  • 8.14000 - 2,3-Dimethoxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
8.14000 EMD Millipore

2,3-Dimethoxybenzoic acid

2,3-Dimethoxybenzoic acid for synthesis. CAS 1521-38-6, chemical formula 2,3-(CH₃O)₂C₆H₃COOH., for synthesis

  • CAS Number 1521-38-6

  • Linear Formula C₉H₁₀O₄

  • Molecular Weight 182.17 g/mol

  •  EC Number 216-188-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8140000010)
solubility   5.310 g/l slightly soluble
mp   121 - 124 °C
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 121 °C
  Melting range (upper value): ≤ 124 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Precautionary Statement(s) 
P260: Do not breathe dust.
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 22-24/25
Do not breathe dust.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?