• USA Home
  • 8.14001 - 3,3-Diethoxypropionitrile

EMAIL THIS PAGE TO A FRIEND
8.14001 EMD Millipore

3,3-Diethoxypropionitrile

3,3-Diethoxypropionitrile for synthesis. CAS No. 2032-34-0, EC Number 217-988-6., for synthesis

Synonym: Cyanoacetaldehyde diethyl acetal, 1,1-Diethoxy-2-cyanoethane

  • CAS Number 2032-34-0

  • Linear Formula C₇H₁₃NO₂

  • Molecular Weight 143.18 g/mol

  •  EC Number 217-988-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   82 °C (13 hPa)
vapor pressure   0.14 hPa (20 °C)
solubility   32 g/l
density   0.95 g/cm3 (20 °C)
flash point   83 °C
autoignition temp.   395 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.949 - 0.952
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 25
Avoid contact with eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?