• USA Home
  • 8.14002 - Tetra-n-octylammonium bromide

EMAIL THIS PAGE TO A FRIEND
8.14002 EMD Millipore

Tetra-n-octylammonium bromide

Tetra-n-octylammonium bromide for synthesis. CAS 14866-33-2, EC Number 238-936-9, chemical formula (C₈H₁₇)₄N⁺Br⁻., for synthesis

  • CAS Number 14866-33-2

  • Linear Formula C₃₂H₆₈BrN

  • Molecular Weight 546.8 g/mol

  •  EC Number 238-936-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8140020010)
solubility   600 g/l
flash point   100 °C
mp   95 - 98 °C
pH   7 (100 g/l, H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 95 °C
  Melting range (upper value): ≤ 98 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2923 90 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?