• USA Home
  • 8.14003 - 2-Chlorophenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.14003 EMD Millipore

2-Chlorophenylacetic acid

2-Chlorophenylacetic acid for synthesis. CAS 2444-36-2, chemical formula 2-(Cl)-C₆H₄CH₂COOH., for synthesis

Synonym: OCPA

  • CAS Number 2444-36-2

  • Linear Formula C₈H₇ClO₂

  • Molecular Weight 170.59 g/mol

  •  EC Number 219-482-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8140030010)
mp   93 - 96 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 93 °C
  Melting range (upper value): ≤ 96 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P260: Do not breathe dust.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?