• USA Home
  • 8.14062 - Allyltrimethylsilane

EMAIL THIS PAGE TO A FRIEND
8.14062 EMD Millipore

Allyltrimethylsilane

Allyltrimethylsilane for synthesis. CAS No. 762-72-1, EC Number 212-104-5., for synthesis

Synonym: 3-Trimethylsilyl-1-propene

  • CAS Number 762-72-1

  • Linear Formula C₆H₁₄Si

  • Molecular Weight 114.26 g/mol

  •  EC Number 212-104-5

Purchase

Properties

packaging   10 ml in Glass bottle (8140620010)
bp   84 - 86 °C
density   0.716 g/cm3 (20 °C)
flash point   -9 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.715 - 0.719
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H225: Highly flammable liquid and vapour.
H315: Causes skin irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P403 + P235: Store in a well-ventilated place. Keep cool.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Flammable  Flammable
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
highly flammable, irritant
R Phrase 
R 11-38
Highly flammable.Irritating to skin.
S Phrase 
S 7-16-26
Keep container tightly closed.Keep away from sources of ignition - No smoking.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?