• USA Home
  • 8.14071 - (1R)-(-)-Camphorquinone

EMAIL THIS PAGE TO A FRIEND
8.14071 EMD Millipore

(1R)-(-)-Camphorquinone

(1R)-(-)-Camphorquinone for synthesis. CAS 10334-26-6, molar mass 166.21 g/mol., for synthesis

Synonym: (1R)-2,3-Bornanedione

  • CAS Number 10334-26-6

  • Linear Formula C₁₀H₁₄O₂

  • Molecular Weight 166.21 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Plastic bottle (8140710001)
mp   198 - 201 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 198 °C
  Melting range (upper value): ≤ 201 °C
  Spec. rotation [α²0/D (c=2 in Chloroform): -106.0 - -102.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?